Econo™ Sterile Webster Needle Holders, Floor Grade, Sklar
Supplier: Sklar
96-2586 96-2587 96-2659
89023-916CS 532.25 CAD
89023-916 89023-918 89023-976
Econo™ Sterile Webster Needle Holders, Floor Grade, Sklar
Forceps
Econo™ Sterile Webster Needle Holders are used with small needles and fine suture material, in plastic surgery or subcutaneous procedures.
They are available with smooth or serrated jaws. Smooth jaws will cause the least damage to the needle; however, serrated jaws hold the needle more tightly. Physician preference determines the type of working surface used.
Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments made in Pakistan. Econo™ Sterile instruments eliminate the need for cleaning and reprocessing. Because they are both sterile and disposable,More Product Information