Surgi-OR™ TC Crile-Wood Needle Holder, Mid Grade, Sklar®
Supplier: Sklar
95-138 95-139
89023-500EA 283.6 CAD
89023-500 89023-502
Surgi-OR™ TC Crile-Wood Needle Holder, Mid Grade, Sklar®
Forceps
The Surgi-OR™ TC Crile-Wood Needle Holder is used to hold and guide small to medium size needles and suture materials.
The Crile-Wood Needle Holder has a gently tapered, blunt tip and is more delicate than the Mayo-Hegar but slightly less delicate than Webster, Halsey, or DeBakey needle holders. The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures.
Hand-crafted in Pakistan, Surgi-OR™ TC mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. They are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. Surgi-OR™ instruments are a cost-effective line of instruments that must meet Sklar's quality standards.