Surgi-OR™ TC Baumgartner Needle Holders, Mid Grade, Sklar®
Supplier: Sklar
95-135 95-832
89023-494EA 242.67 CAD
89023-494 89023-730
Surgi-OR™ TC Baumgartner Needle Holders, Mid Grade, Sklar®
Forceps
Surgi-OR™TC Baumgartner Needle Holders feature heavy serrations which make this instrument capable of firmly holding larger needles and dense suture material
Hand-crafted in Pakistan, Surgi-OR™ TC mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. They are designed to stay sharp for a longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
They are used in procedures on the trunk and limbs.