VWR® Serological Pipette
Supplier: VWR International
VWR®
56700-5110
53283-490CS 1289.43 CAD
53283-490
VWR® Serological Pipette
Pipettes Serological Pipettes
These serological plastic pipettes are designed to use with tissue culture work.
  • Black printed scale
  • Calibrated to deliver to the tip, with two rings near the top
  • Irradiation sterilized