Magid® EconoWear® Disposable Laboratory Coats, Magid
Supplier: Magid Glove
CC1114M CC1114L CC1114XL CC1114XXL CC1114XXXL
13450-156CS 2866.01 CAD
13450-156 13450-158 13450-160 13450-162 13450-164
Magid® EconoWear® Disposable Laboratory Coats, Magid
Disposable Garments
EconoWear® disposable lab coats are made of Tyvek, a tough polyolefin material, which provides the optimal balance of protection, durability and comfort.
  • Elastic wrists
  • Collar with snap front
  • Greater range of movement while stretching and bending
These coveralls feature a great range of movement while stretching and bending, improved mobility, a tailored fit, and reinforcement in high stress areas. Elastic at wrist openings creates a comfortable seal against contaminants. For use against such non-hazardous materials as dirt and grime, animal waste,More Product Information