Sklarlite™ Metzenbaum Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
23-1175 23-1172 23-1173 23-1174 23-1180 23-1182
10194-216EA 382.83 CAD
10194-216 10195-094 10195-096 10195-098 10195-108 10195-110
Sklarlite™ Metzenbaum Dissecting Scissors, OR Grade, Sklar
Scissors
Sklarlite™ Metzenbaum Dissecting Scissors are used for dissecting and cutting tissue only
Sklarlite™ instruments are a lighter version of standard Sklar instruments. A manufacturing process developed by Sklar shaves microns off standard and delicate pattern instruments, while maintaining structural integrity and durability. This lightweight modification provides the surgeon with greater control and less fatigue.
These scissors are not recommended for cutting sutures, drains, or heavy tissue.
Order Now
SPECIFICATIONS
Environmentally Preferable