Sklarlite XD™ Mayo Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
23-1168 23-1169 23-1170 23-1171
10195-086EA 314.65 CAD
10195-086 10195-088 10195-090 10195-092
Sklarlite XD™ Mayo Dissecting Scissors, OR Grade, Sklar
Scissors
Sklarlite XD™ Mayo Dissecting Scissors are an extra light and extra delicate version of the standard Mayo scissors that offer the surgeon the highest degree of precision with the least amount of fatigue, while maintaining structural integrity and strength
While the curved instrument is used for cutting or dissecting dense or deep tissue, and the straight version is used for cutting surface tissue or sutures, both may be used depending on procedure and physician preference.
Mayo Dissecting Scissors are a versatile instrument, that can be used in multiple procedures and settings.
Ordering information: Available straight or curved in a variety of lengths with a standard beveled blade.