Vibration Dampening Mounts, METTLER TOLEDO®
Supplier: Mettler Toledo
63053978 30238400 30238401
CA11277-542EA 965.33 CAD
CA11277-542 76492-552 76492-554
Vibration Dampening Mounts, METTLER TOLEDO®
Furniture Tables and Workbenches
Vibrations can negatively influence weighing results. A proper balance table or marble slab is necessary to ensure ergonomic and efficient weighing in a lab.
  • Speeds up the working process
  • Increases the quality of results
Anti-vibration tables enable proper balance operation and faster stabilization times.