KIMBLE® Safety Tip Needles with Blunt Ends, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
868280-2001 868280-1801 868280-2201
KT868280-2001PK 14.48 CAD
KT868280-2001 KT868280-1801 KT868280-2201
KIMBLE® Safety Tip Needles with Blunt Ends, DWK Life Sciences
Syringe Needles
Blunt end safety tip needles are used for general purpose dispensing in laboratory applications.
  • Nonsterile stainless steel needle with an inert plastic luer hub
  • Blunt end for safety