Sklarlite™ Kelly Hemostatic Forceps, OR Grade, Sklar
Supplier: Sklar
23-2107 23-2108
10192-348EA 225.24 CAD
10192-348 10192-472
Sklarlite™ Kelly Hemostatic Forceps, OR Grade, Sklar
Forceps
Sklarlite™ Kelly Hemostatic Forceps are used for clamping large blood vessels or manipulating heavy tissue
They are available curved or straight with a ratcheted finger ring handle. Similar to Rankin forceps, the jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws, however, Kelly forceps are shorter and heavier than Rankin forceps. Sklarlite™ instruments are a lighter version of standard Sklar instruments. This lightweight modification provides the surgeon with greater control and less fatigue.
They may also be used for soft tissue dissection.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.