Canners Vacuum Guage, Jensen Instrument
Supplier: Jensen Tools + Supply
50462
31757-128EA 1313.68 CAD
31757-128
Canners Vacuum Guage, Jensen Instrument
Pumps Vacuum Gauges
Canners Gauge is used for testing vacuum of packaged product such as canned food or drink.
  • Easy to use for destructive sampling tests
  • 1" graduations in Hg, inside 2" diameter case
  • Tip can be cleaned for multiple use, non-removable
  • The rubber gasket prevents air from entering the container during the test
  • Force the stainless steel needle through the top of the container while reading the gauge