Silver sulfate ACS, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Disilver sulphateSilver(I) sulfateDisilver sulfate

BDHVBDH4560-100G
BDH4560-100GEA 1300.05 CAD
BDH4560-100G
Silver sulfate ACS, VWR Chemicals BDH®
Silver sulphate

Powder.

Formula: Ag₂SO₄
MW: 311.8 g/mol
Boiling Pt: 1085 °C (1013 hPa)
Melting Pt: 655 °C
Density: 5.45 g/cm³ (20 °C)
MDL Number: MFCD00003407
CAS Number: 10294-26-5
Merck Index: 13,08603
https://ca.vwr.com/stibo/web/std.lang.all/94/57/2299457.jpg

Order Now

Specification Test Results

Assay Min. 98.0 %
Insoluble matter and silver chloride Max. 0.02 %
Substances not precipitated by HCl Max. 0.03 %
NO3 (Nitrate) Max. 0.001 %
Fe (Iron) Max. 0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR