VWR® DURAC® Safety Specific Gravity/Relative Density (g/cm³) Combined Form Thermo-Hydrometers, Traceable to NIST
Supplier: Avantor
61821-0511 61821-0211 61821-0411 61821-0311
30617-762EA 150.53 CAD
30617-762 30617-764 30617-760 30617-766
VWR® DURAC® Safety Specific Gravity/Relative Density (g/cm³) Combined Form Thermo-Hydrometers, Traceable to NIST
Hydrometers
Made of Lead free ballast and glass, these combined form thermo-hydrometers offer the additional convenience of an enclosed thermometer with a temperature scale of 0/150°F, and measure specific gravity / relative density (g/cm3) of a variety of liquids in various ranges from 0.800 to 1.420
  • Conform to the accuracy requirements for ASTM thermo-hydrometers
  • Commonly used in industries such as food and beverage, chemical, pharmacology, transportation, refrigeration and more
  • 381mm (15”) long
  • Lead free ballast and glass
  • Temperature standardization: 60°F (15°C)
  • Thermometer contains mercury-free blue spirit organic liquid fill
  • Individually serialized
Certifications: DURAC® Safety Combined Form Thermo-Hydrometers are hand inspected by our U.S. quality team in our exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC /MRA and located in our ISO 14001:2004 Environmental Management System registered facility.
Ordering information: Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.
Caution: Combined form hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures. Enclosed thermometers should be used as a general reference only. For temperature critical applications, a calibrated standard thermometer is recommended.