Criterion™ Pre-Weighed Dehydrated Culture Media in a Mylar® Pouch, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5000 C6530 C6550 C6650 C7430 C8930 C8940 C8950
CA89405-064EA 218.29 CAD
CA89405-064 CA89405-832 CA89405-848 CA89405-920 CA89406-340 CA89406-828 CA89406-836 CA89406-844
Criterion™ Pre-Weighed Dehydrated Culture Media in a Mylar® Pouch, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
Mylar® Zip-bags contain a pre-weighed amount of dehydrated culture media sufficient to prepare two liters of finished product (after water is added). Criterion™ is designed to meet or exceed the highest standards.
  • Convenient pouch
  • Easy to use
  • Efficient storage
  • Pre-measured