Pure Gum Rubber Tubing
Supplier: PRINCE RUBBER & PLASTICS CO.
PG40-0188-0063 PG40-0500-0215 PG45-0188-0313 TPUR-0375-0500 PG45-0500-0125 PG45-0125-0063
470334-504EA 565.05 CAD
470334-504 470334-506 470334-510 470334-512 470334-514 470334-508
Pure Gum Rubber Tubing
Tubing Transfer Tubing
This tubing can be mandrel coated or have a smooth finish.
  • Mandrel-coated tubing: 40 ±5 (Shore A) Durometer
  • Smooth finish tubing: 45 ±5 (Shore A) Durometer