Prefilled Zinc Formalin Containers, Azer Scientific
Supplier: Azer Scientific
PFZF-020 PFZF-040 PFZF-060 PFZF-090 PFZF-120
CA10813-594CS 1113.19 CAD
CA10813-594 CA10813-596 CA10813-622 CA10813-624 CA10813-626
Prefilled Zinc Formalin Containers, Azer Scientific
Sample Containers General Purpose Containers
Prefilled containers, half filled with zinc formalin fixative, are ideal for all tissue types. They promote excellent H&E staining and enhance IHC results. Also enhances nuclear and cytoplasmic fixation.
  • Click-Top Closure
  • Optimal for All Tissue Types
  • Chipboard Material Packaging
Containers are packaged in convenient transport packs which allow for easy distribution to other labs and doctor's offices
Containers feature a click-top closure to ensure the user that the seal is fully engaged.
The packs are made of chipboard material and are safe for operating room use.
Certifications: All containers sized 20 to 120mL are also 95kPa compliant for safe use in pneumatic tube systems.