Ampicillin sodium salt

Supplier: G-Biosciences

Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt

RC-020 RC-021
CA71003-352EA 565.01 CAD
CA71003-352 CA71003-354
Ampicillin sodium salt
Ampicillin sodium salt

Ampicillin is an inhibitor of bacterial cell wall synthesis which acts by inhibiting the cross-linking of peptidoglycan


It is used in the selection of antibiotic cell hybrids, and has a high rate of action against many gram negative bacterial strains.

Formula: C₁₆H₁₈N₃NaO₄S
MDL Number: MFCD00064313
CAS Number: 69-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR