SpillEx Absorbent Floor Pads, Vileda Professional-FHP
Supplier: Vileda Professional - FHP
146760 150738 150739
10756-160CS 1794.06 CAD
10756-160 10841-568 10841-570
SpillEx Absorbent Floor Pads, Vileda Professional-FHP
Spill Control Materials
SpillEx Pads are the ideal solution for spill pickup or equipment leaking, makes free from the risk of cross contamination.
  • Highly absorbent pads are ideal for removal of water based spills
  • Ultra-thin construction moves excess liquid quickly to absorbent core
  • Water-impermeable film on back side allows no-touch removal after use
  • High concentration of super absorbent particles for holding high volume of liquid without dripping
  • Will not drip when lifting and moving, as SpillEx transforms liquid to a gel
  • Ideal for picking up a spillage already on the floor or for use to collect dripping from equipment above the floor
Absorb spills quickly with SpillEx Absorbent Floor Pads
Standard SpillEx Pads are ideal for use in laboratory, LAR, production, and ISO 7 enivronments. Cleanroom SpillEx Pads are manufactured with an extra-strong film, and are ideal for use inside Core Cleanrooms up to ISO 5. Cleanroom SpillEx Pads are supplied vacuum sealed in a poly double bag with an easy open notch. The gamma irradiated pads feature a gamma processed indicator dot.
SpillEx is a disposable, super-absorbing floor cloth that takes care of liquid spills in minutes. When tested against leading competitive products, SpillEx shows the lowest particle count. One SpillEx absorbs and holds up to 1,200mL of water and up to 500mL of NsCl 0.9% (similar to urine). Pads will not drip when lifting, as SpillEx transforms the liquid into a gel.