SGE Syringes, General Purpose Manual Syringe, NanoVolume, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
000303 000350 000353 000355 000380 000501 000506 000510 000550 000553 000800 000801 000803 000804 000850 000852 000300 000310 000311 000500 000802 000301 000376 000570
14225-100EA 257.29 CAD
14225-100 14225-102 14225-104 14225-106 14225-110 14225-114 14225-116 14225-120 14225-122 14225-124 14225-128 14225-130 14225-132 14225-134 14225-138 14225-140 60361-000 60361-004 60361-006 60361-022 60361-024 60361-034 60361-414 60361-416 60361-418
SGE Syringes, General Purpose Manual Syringe, NanoVolume, Trajan Scientific and Medical
Syringes Syringes for Chromatography
With the ability to inject down to 50nL with high precision and accuracy, SGE NanoVolume Plunger-in-Needle Syringes are perfect for nanovolume capillary chromatography injection.
  • Removable needle
  • Borosilicate glass barrel
  • Accuracy and reproducibility ±2%
The sample is only drawn up into the needle, not the syringe barrel. When the plunger is depressed, the sample is completely dispensed by the NanoVolume plunger that extends right to the tip of the needle.