Tumor Grasping Forceps, OR Grade, Sklar
Supplier: Sklar
98-7758 98-7772 98-7773 98-7720
10185-180EA 2715.41 CAD
10185-180 75995-568 75995-570 75996-294
Tumor Grasping Forceps, OR Grade, Sklar
Forceps
Tumor grasping forceps vary in overall length, tip configuration, and tip diameter.
  • Reusable
  • Nonsterile
The bayonet style tumor grasping forceps are commonly used in neurosurgical procedures involving tumor removal. The slim bayonet handle eases access to small delicate spaces during craniotomy procedures.