TC Jacobson Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-8055 21-8058 21-8059 21-8049 21-8052 21-8053 21-8054 21-8056 21-8057
89023-386EA 1837.89 CAD
89023-386 89023-388 10185-276 76192-664 76192-718 76192-720 76192-722 76192-724 76192-726
TC Jacobson Needle Holder, OR Grade, Sklar®
Forceps
Sklar's® Jacobson Tungsten Carbide Microvascular Needle Holder is primarily used for suturing in microvascular and cardiovascular procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Also known as a Jacobson-Castroviejo Needle Holder, the Jacobson Tungsten Carbide Microvascular Needle Holder features finely cross-serrated, tapered jaws that are able to securely hold very fine sutures. Sklar® permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. Serrations on TC instruments are designed to stay sharp for a significantly longer period of time than standard instruments.