TC Castroviejo Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-8040 21-8042 21-8043 21-8044 21-8045 21-8046 21-8047 21-8048
89023-374EA 1876.55 CAD
89023-374 89023-376 89023-378 89023-380 89023-382 89023-384 76192-660 76192-662
TC Castroviejo Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® Tungsten Carbide Castroviejo Needle Holder is a multipurpose microsurgical instrument that is often used in ophthalmic and plastic surgeries.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Castroviejo Needle Holder features a double spring, flat handle, and it is available with straight or curved jaws. It is also available with or without a locking mechanism. The small tips of this needle holder are ideal for handling small, delicate needles and sutures. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar® permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles.