Graves Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
90-3803 90-3701 90-3702 90-3700 90-3802 90-3800
10192-440EA 321.69 CAD
10192-440 10193-512 10193-732 10193-760 10193-770 10197-214
Graves Vaginal Speculum, OR Grade, Sklar
Specula
Graves Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, Pap Smears, and examining the cervix.
It is a double blade duckbill speculum inserted with the blades closed.
Censitrac Ready products are industry standard and constructed of stainless steel.
The blades are then opened by an external lever and screw.The Graves Speculum has wider blades than the Pederson Speculum and the practitioner can best determine, which type and size speculum is most appropriate for the patient.
Ordering information: Censitrac instruments are custom barcoded toMore Product Information