TC Crile-Wood Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-8006 21-8007 21-8008 21-8009 21-8010 21-8011 21-4914 21-8003
89023-316EA 456.09 CAD
89023-316 89023-318 89023-320 89023-322 89023-324 89023-326 76192-544 76192-648
TC Crile-Wood Needle Holder, OR Grade, Sklar®
Forceps
TC Crile-Wood Needle Holder is used to hold and guide small to medium size needles and suture materials.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Crile-Wood Needle Holder has a gently tapered, blunt tip and is more delicate than the Mayo-Hegar but less delicate than Webster, Halsey, or DeBakey needle holders. The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.