TC Webster Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-8002 21-8080 21-2240 21-2241
89023-310EA 453.74 CAD
89023-310 89023-404 76192-386 76192-388
TC Webster Needle Holder, OR Grade, Sklar®
Forceps
Sklar's® TC Webster Needle Holder is used with small needles and fine suture material, commonly in plastic surgery or subcutaneous procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Smooth jaws will cause the least damage to the needle, however, serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. Tungsten Carbide instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar® permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.