Pierce 3UV Lamp, Thermo Scientific
Supplier: Thermo Scientific
95034 95035
CAPI95034EA 2167.62 CAD
CAPI95034 CAPI95035
Pierce 3UV Lamp, Thermo Scientific
Lights UV Lights, Lamps and Bulbs
Thermo Scientific Pierce UV Lamps are compact, multifunctional ultraviolet lamps with 252 nm, 302 nm and 365 nm dial-selectable wavelength settings especially for use with photoactivatable crosslinkers and UV-crosslinking methods.
  • Unique reflector design provides maximum UV for all applications
  • Designed for hand-held or stationary use
  • Lamp features aluminum construction and scratch-resistant finish
  • 8-watt lamps are ideal for a wide area of illumination, such as culture plates
  • This lamp is validated for use in crosslinking applications with diazirine and phenyl azide photoactivatable crosslinkers
15.5 L x 3 W x 4.5" D (394 x 76 x 114 mm).
Caution: For Research Use Only. Not for use in diagnostic procedures.