Pierce™ TMT10plex™ Isobaric Mass Tag Labelling Kit and Reagent Sets, Thermo Scientific
Supplier: Thermo Scientific
TMT10plex™ Isobaric Mass Tag Labeling Reagents Sets enable multiplexed protein identification and quantitative analysis by tandem mass spectrometry (MS).
- Increased sample multiplexing for relative quantitation
- Increased sample throughput
- Fewer missing quantitative channels among samples
Each mass-tagging reagent within a set has the same nominal mass (ie
The TMT10plex™ Label Reagents share an identical structure with Thermo Scientific™ TMTzero™ and TMTsixplex™ Reagents but contain different numbers and combinations of 13C and 15N isotopes in the mass reporter. The different isotopes result in a 10plex set of tags that have monoisotopic mass differences in the reporter that can be detected using high resolution mass spectrometry.
isobaric) and chemical structure composed of an amine-reactive NHS-ester group, a spacer arm and a mass reporter. The reagent set can be used to label up to ten different peptide samples prepared from cells or tissues. For each sample, a unique reporter mass (ie. TMT10 126-131Da) in the low mass region of the MS/MS spectrum is used to measure relative protein expression levels during peptide fragmentation.
Delivery information: Includes reagents sufficient for: One 10-plex (1 x 10-way) experiment, Label reagents: TMT10-126, 1 x 0.8 mg, TMT10-127N, 1 x 0.8 mg, TMT10-127C, 1 x 0.8 mg, TMT10-128N, 1 x 0.8 mg, TMT10-128C, 1 x 0.8 mg, TMT10-129N, 1 x 0.8 mg, TMT10-129C, 1 x 0.8 mg, TMT10-130N, 1 x 0.8 mg, TMT10-130C, 1 x 0.8 mg and TMT10-131, 1 x 0.8 mg.
Caution: For Research Use Only. Not for use in diagnostic procedures.