Pierce™ HisPur™ Affinity Purification Kits, Thermo Scientific
Supplier: Thermo Scientific
89967 90090 89964 89965 89966 89968 89969 90091 90092
CAPI89967PK 634.71 CAD
CAPI89967 CAPI90090 CAPI89964 CAPI89965 CAPI89966 CAPI89968 CAPI89969 CAPI90091 CAPI90092
Pierce™ HisPur™ Affinity Purification Kits, Thermo Scientific
Protein Purification Protein Purification Kits
Pierce™ HisPur™ Cobalt Purification Kit is designed for the efficient purification of recombinant polyhistidine-tagged proteins from bacterial, mammalian and baculovirus-infected cells.
  • Obtain <10 mg of pure His-tagged protein per mL of resin
  • Resin can be reused several times or discarded after single use
  • Cobalt-chelate coordination core binds fewer host protein contaminants
  • Purify proteins under native or denaturing conditions
His-tagged proteins are purified from total soluble protein extract using a cobalt-charged tetradentate chelator immobilized onto 6% crosslinked agarose. Many immobilized metal affinity chromatography (IMAC) resins contain nickel (Ni2+) for purifying His-tagged proteins. Although Ni2+ chelate resins achieve high protein yields, purity is often sub-optimal, requiring additional clean-up steps. Cobalt achieves both high yield and purity with minimal optimisation. The HisPur™ Cobalt Resin binds fewer non specific proteins, displays less metal leaching and enables less stringent elution conditions compared to Ni2+ resins.
Delivery information: Complete kit contains HisPur™ Cobalt spin columns, pre-formulated Equilibration/Wash and Elution Buffers, collection tubes and tip plugs. Resin and spin columns are also available separately.
Caution: For Research Use Only. Not for use in diagnostic procedures.