Pierce™ Zeba™ Desalting Chromatography Cartridges, Thermo Scientific

Supplier: Thermo Scientific
89934 89935
CAPI89934EA 505.21 CAD
CAPI89934 CAPI89935
Pierce™ Zeba™ Desalting Chromatography Cartridges, Thermo Scientific
Chromatography Columns
Zeba™ Desalting Chromatography Cartridges, 7K MWCO, 2 ml contains Zeba™ High-Performance Resin for desalting or buffer exchange applications. Store at 4 to 8 °C.

  • High recovery: Low-binding resin maximizes protein recovery
  • Fast: No fraction screening or waiting for protein to emerge by gravity-flow
  • Easy-to-use: No cumbersome column preparation or equilibration
  • Flexible: Available in spin columns, filter spin plates and cartridges for a range of needs
  • Economical: Great performance at lower cost than other commercially available cartridges

Zeba Desalting Cartridge-fittings are convenient, ready-to-use devices compatible with the major automated liquid-chromatography systems or for manual syringe processing.

Available formats:
Spin Columns - 75 µl, 0.5 ml (microcentrifuge), 2, 5, 10 ml columns
Chromatography cartridges—compatible with automated liquid chromatography systems and with syringe processing

The cartridges attach directly to ÄKTA™ or FPLC Systems without additional connectors. An accessory pack included with each product readily adapts cartridges for use with luer syringe fittings or ¹/₁₆" tubing. Pierce Desalting Cartridges provide fast, easy and reproducible chromatographic separations and can be regenerated for multiple uses and efficiently process samples from 50 to 1500 µl.

Delivery information: Each column pack contains one accessory pack with one female Luer-Lok® adapter, one connector fitting, one column plug and five bottom caps.

Caution: For research use only. Not for use in diagnostic procedures.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR