Pierce™ Affinity Chromatography Cartridges, Protein A/G, Thermo Scientific
Supplier: Thermo Scientific
89930 89931
CAPI89930EA 654.52 CAD
CAPI89930 CAPI89931
Pierce™ Affinity Chromatography Cartridges, Protein A/G, Thermo Scientific
Chromatography Columns
The Thermo Scientific™ Pierce™ Chromatography Cartridges Protein A/G are convenient, ready-to-use prepacked devices for isolation and purification of most immunoglobins.
  • Protein A/G: Immobilized recombinant fusion protein of the antibody-binding domains of Protein A and Protein G enables polyclonal IgG purification from nearly any mammalian species
  • Agarose resin: Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Inert and stable: Superior manufacturing method immobilizes Protein A/G by charge-
More Product Information