Pierce™ Affinity Chromatography Cartridges, Protein G, Thermo Scientific
Supplier: Thermo Scientific
89926 89927
CAPI89926EA 616.11 CAD
CAPI89926 CAPI89927
Pierce™ Affinity Chromatography Cartridges, Protein G, Thermo Scientific
Chromatography Columns
These cartridges are convenient, ready-to-use prepacked devices for isolation and purification of mammalian polyclonal and monoclonal antibodies from serum, ascites and cell culture supernatants.
  • Protein G: Immobilized Protein G is ideal for polyclonal IgG purification from mouse, human, cow, goat and sheep serum, including human IgG3 and mouse IgG1 isotypes
  • Agarose resin: Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Inert and stable: Superior manufacturing method immobilizes Protein G by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield
  • Standard capacity: Pierce Protein G Agarose has a normal load of immobilized Protein G, providing a binding capacity of 11 to 15 mg human IgG/ml resin
  • Storage temperature 4 to 8 ˚C
These cartridges are compatible with the major automated liquid-chromatography systems or for manual syringe processing, and attach directly to ÄKTA™ or FPLC Systems without additional connectors.
An accessory pack, included with each product, readily adapts columns for use with Luer-Lok Syringe Fittings or ¹/₁₆" tubing. The Protein G cartridges provide fast, easy and reproducible chromatographic separations and can be regenerated for multiple uses.
Delivery information: Kit includes an accessory pack (1 female Luer-Lok® adapter, 1 connector fitting, 1 column plug and bottom caps).
Caution: For research use only, not for use in diagnostic procedures.