Pierce™ Affinity Chromatography Cartridges, Protein A, Thermo Scientific
Supplier: Thermo Scientific
89924 89925
CAPI89924EA 532.15 CAD
CAPI89924 CAPI89925
Pierce™ Affinity Chromatography Cartridges, Protein A, Thermo Scientific
Chromatography Columns
These cartridges are convenient, ready to use pre-packed devices for the isolation and purification of polyclonal and monoclonal antibodies. These cartridges are effective for routine affinity purification of polyclonal and monoclonal IgG from serum, ascites and cell culture supernatant.
  • Immobilized Protein A is ideal for polyclonal IgG purification from human, rabbit, pig, dog or cat serum
  • Resin support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Binding capacity >34 mg human IgG/mL resin (approx. 16 to 17 mg mouse IgG/ml resin)
  • Upon receipt do not freeze the product
  • Storage temperature 4 to 8 ˚C
The cartridges are compatible with the major automated liquid-chromatography systems or manual syringe processing
The included accessory pack readily adapts cartridges for use with Luer-Lok® syringe fittings or ¹/₁₆" tubing. These cartridges enable fast, easy and reproducible chromatographic separations and can be regenerated for multiple uses.
The cartridges attach directly to ÄKTA™ or FPLC Systems without additional connectors.
Delivery information: Kit includes an accessory pack (1 female Luer-Lok® adapter, 1 connector fitting, 1 column plug and bottom caps).
Caution: For research use only, not for use in diagnostic procedures.