Pierce™ NHS-Azide and NHS-Phosphine Reagents, Thermo Scientific

Supplier: Thermo Scientific

26130 88900 88902
CAPI26130EA 1142.73 CAD
CAPI26130 CAPI88900 CAPI88902
Pierce™ NHS-Azide and NHS-Phosphine Reagents, Thermo Scientific
Protein Linking and Labeling Systems

The NHS-Azide and NHS-Phosphine Reagents are amine-reactive compounds for derivatising primary amines of proteins or amine-coated polymer surfaces. Once a protein or surface is azide- or phosphine-labelled, the two components are mixed for effective and stable conjugation. Phosphine groups react with azides via a Staudinger reaction to produce an aza-ylide intermediate that is trapped to form a stable, covalent amide bond. Because phosphines and azides are absent from biological systems, there is minimal background labelling of macromolecules found in cells or lysates.


  • Reagent Easily Dissolves in Water-Miscible Solvents for Subsequent Dilution in Aqueous Reaction Mixtures with Cell Lysates and Other Biological Samples
  • Compatible Reaction Chemistry Occurs Effectively in Simple Buffer Conditions; Requires no Accessory Reagents Such as Copper or Reducing Agents
  • NHS Esters are Specific for Covalent Attachment to Primary Amines (Side Chain of Lysine or N-terminus of Polypeptides)
  • Chemoselective Azide and Phosphine Groups do not React or Interfere with Components of Biological Samples (Bioorthogonal) but Conjugate to One Another with High Efficiency


This NHS-ester compound reacts to form covalent bonds with primary amines (e.g., side chain of lysine residues or aminosilane-coated surfaces). The azide (N3) group reacts with phosphine-labeled molecules by a mechanism known as Staudinger chemistry, enabling efficient and specific conjugation of derivatized molecules in biological samples.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now

SPECIFICATIONS

Environmentally Preferable
Host
Shipping temperature ambient
Storage conditions Upon receipt store at –20 ˚C protected from light and moisture

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR