Pierce™ Protein L Magnetic Beads, Thermo Scientific
Supplier: Thermo Scientific
88849 88850
CAPI88849EA 653.62 CAD
CAPI88849 CAPI88850
Pierce™ Protein L Magnetic Beads, Thermo Scientific
Beads Magnetic Beads
Thermo Scientific Pierce Protein L Magnetic Beads are high-capacity and high-throughput affinity particles for antibody purification using manual or robotic magnetic separators.
  • Immobilized Protein L is Ideal for Selective Purification of Human and Mouse Antibodies that have kappa Light Chains
  • Low Non-Specific Binding of Stable, Pre-Blocked Beads Provide Clean Purification of Antibody
  • Beads are Compatible with Manual and Automated Workflows Making it Versatile
Recombinant Protein L, covalently attached to a blocked magnetic bead surface, selectivelyMore Product Information