Pierce™ Protein Magnetic Beads, Protein A, Protein G, Thermo Scientific
Supplier: Thermo Scientific
88845 88846 88847 88848
CAPI88845EA 590.92 CAD
CAPI88845 CAPI88846 CAPI88847 CAPI88848
Pierce™ Protein Magnetic Beads, Protein A, Protein G, Thermo Scientific
Beads Magnetic Beads
Thermo Scientific Pierce protein magnetic beads are high-capacity and high-throughput affinity particles for antibody purification and immunoprecipitation methods using manual or robotic magnetic separators.
  • Higher efficiency in yield of IP target antigens
  • Low non-specific binding of stable, pre-blocked beads provide highly purified product
  • Magnetic beads eliminate resin loss and provide for more efficient separation than traditional IP methods that use only centrifugation
  • Beads are compatible with manual and automated workflows making it versatile
Recombinant protein A, covalently attached to a blocked magnetic bead surface, can bind to antibodies from many different species, enabling purification of antibodies from crude extracts
These beads are used for purifying antibody from serum, cell culture supernatant or ascites, as well as for IP/Co-IP of antigens from cell or tissue extracts. Protein A can bind to antibodies from many different species, including mouse, human, rabbit, pig, dog, and cat, while Protein G binds to antibodies from mouse, human, rabbit, cow, goat and sheep.They can be used manually with a magnetic stand, as well as with automated platforms. Antibody or antigen/antibody complex (IP) is first captured on the magnetic beads. The beads are washed and then the target is eluted with low pH elution buffer.
Immunoprecipitation assays performed with protein A coated beads result in high yield of target antigen with very low background. Using our crosslinker chemistry, you can immobilize an antibody onto the magnetic particle and prevent IgG contamination in IP or Co-IP experiments. The Pierce Protein A Magnetic Beads can be used manually with a magnetic stand, as well as with automated platforms, such as the Thermo Scientific KingFisher Instruments, for high-throughput workflows.
Delivery information: Upon receipt store at 4?C. Product shipped with an ice pack.
Caution: For Research Use Only. Not for use in diagnostic procedures.