Pierce™ Anti-c-Myc Magnetic Beads
Supplier: Thermo Scientific
88842 88843
CAPI88842EA 1229.52 CAD
CAPI88842 CAPI88843
Pierce™ Anti-c-Myc Magnetic Beads
Beads Magnetic Beads
Thermo Scientific Pierce Anti-c-Myc Magnetic Beads are affinity particles for immunoprecipitation of recombinant c-Myc-tagged proteins expressed in bacterial or mammalian cells or in vitro systems, using manual or robotic magnetic separators.
  • Highly Specific anti-c-Myc Monoclonal Antibody Enables High Yield and High Purity Immunoprecipitation
  • Fast and Convenient with Low Non-Specific Binding
  • Beads are Compatible with Manual and Automated Workflows
The blocked magnetic bead surface is coated with anti-c-Myc antibody, a highly specific mouse IgG1 monoclonal antibody (clone 9E10) that recognizes the c-Myc-epitope tag (EQKLISEEDL) derived from the human c-myc oncogene (p62 c-myc)
Pierce Anti-c-Myc Magnetic Beads are convenient for the immunoprecipitation (IP) of recombinant c-Myc tagged proteins and the co-immunoprecipitation (Co-IP) of their interacting proteins. The beads are incubated with a cell lysate containing c-Myc tagged protein and the fusion protein is captured. The beads are subsequently washed and then the target proteins are eluted using 0.1M glycine (pH 2.0) 50 mM NaOH, or SDS-PAGE sample buffer, the protocol has been optimized for each of these conditions. Anti-c-Myc antibody can be used to detect c-Myc tagged protein by Western blot analysis.
The Pierce Anti-c-Myc Magnetic Beads can be used manually with a magnetic stand, as well as with automated platforms, such as the Thermo Scientific KingFisher Instruments, for high-throughput workflows.
Ordering information: Store at 4 °C.
Caution: For Research Use Only. Not for use in diagnostic procedures.