Pierce™ Streptavidin Magnetic Beads, Thermo Scientific
Supplier: Thermo Scientific
88816 88817
CAPI88816EA 874.48 CAD
CAPI88816 CAPI88817
Pierce™ Streptavidin Magnetic Beads, Thermo Scientific
Beads Magnetic Beads
Thermo Scientific Pierce Streptavidin Magnetic Beads accelerate throughput for automated magnetic purification of biotinylated molecules.
  • High-Performance Beads—non-aggregating, Pre-Blocked, Iron Oxide, Superparamagnetic Microparticles Provide Exceptional Uniformity for Automated HTS and Manual Applications Alike
  • Stable Immobilization Chemistry—Streptavidin is Immobilized using Leach-Resistant Chemistry
  • High Capacity—Superior Quality Beads with High Binding Capacity Provide Rapid and Efficient Biomolecule Purification from Complex Samples
  • Low Non-Specific
More Product Information