Pierce™ ESI Ion Calibration Solution, LTQ, Thermo Scientific
Supplier: Thermo Scientific
88322 88323
CAPI88322EA 716.17 CAD
CAPI88322 CAPI88323
Pierce™ ESI Ion Calibration Solution, LTQ, Thermo Scientific
Standards Colour Reference and Spectroscopy Standards
The Thermo Scientific Pierce LTQ ESI Positive Ion Calibration Solution is a mixture of highly purified ionizable molecules specifically designed for positive mode calibration of Thermo Scientific ion trap and orbitrap instruments.
  • Strong peaks mixture of caffeine, MRFA and Ultramark 1621 in a acetonitrile/methanol/acetic solution.
  • Ready-to-use load the mass reference standard into a syringe and inject into the instrument
  • High-purity mass spectrometry grade reagents in a non-leachable container
  • Stable store at room temperature for more than one year
The Pierce LTQ ESI Positive Ion Calibration Solution is a ready-to-use liquid formulation ideal for quickly performing the routine calibration required to maintain the robust performance of Thermo Scientific Mass Spectrometers
The LTQ ESI Positive Ion Calibration Solution can be used to calibrate the LTQ series, the LTQ Orbitrap series, the LXQ, LCQ FLEET and the Exactive Mass Spectrometer instruments.
The LTQ ESI Positive Ion Calibration Solution is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications. The stable solution is provided in a leak-proof, high-purity PTFE bottle.
Caution: For Research Use Only. Not for use in diagnostic procedures.