Pierce™ Mass Spectrometry Controls, Peptide Retention Time Calibration Mixture, Thermo Scientific
Supplier: Thermo Scientific
Pierce peptide retention time calibration mixture enables reversed phase liquid chromatography (LC) and mass spectrometry (LC-MS) users to optimize and confirm correct operation of their equipment.
- Assess chromatography and MS instrument performance
- Predict peptide retention across multiple instrument platforms
- Predict peptide retention time from sequence using calculated hydrophobicity factor
- Optimize scheduled MS acquisition windows for improved quantification and increased multiplexing
- Normalize collision energies between instruments and platforms
- Normalize results for variation in retention times and peak intensities between runs
Testing the mixture frequently throughout the lifetime of a column or including it in quantitative samples makes it possible to monitor and normalise LC-MS performance between samples and over time.
Caution: For Research use only. Not for use in diagnostic procedures.