Pierce™ Detergent Removal Spin Plates, Thermo Scientific

Supplier: Thermo Scientific
Pierce™
88304 88304
CAPI88304EA 2023.76 CAD
CAPI88304 PI88304
Pierce™ Detergent Removal Spin Plates, Thermo Scientific
Microplates Filter Plates
Pierce™ Detergent removal filter spin plates enable multiple-sample processing using a high-performance resin that offers effective detergent removal and excellent protein/peptide recovery.

  • Fast and efficient protocol
  • Removes greater than 95% of detergents with high protein recovery
  • Helps improves mass spectrometry peptide coverage
  • Best for processing samples with proteins or peptides at greater than 100 µg/ml

The Pierce™ Detergent removal resin specifically binds a wide variety of detergents and surfactants that are commonly used in protein extraction and biological sample preparation and provide an especially convenient and rapid format for treating protein and peptide solutions to remove interfering detergents before downstream analysis by mass spectrometry and other techniques.

Pierce™ Detergent removal resin efficiently binds to and removes high concentrations of detergents from 10 µl to 1 ml samples with minimal sample loss.

Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR