Pierce™ High Capacity Endotoxin Removal Resin, Thermo Scientific
Supplier: Thermo Scientific
Pierce™ High Capacity Endotoxin Removal Resins are ideal for the removal of endotoxins from protein or antibody samples prior to cell transfection or animal injection.
- Bind up to 2 000 000 EU/ml to eliminate <99% of endotoxins
- Recover greater than 85% of protein sample
Pierce™ High Capacity Endotoxin Removal Resin is a porous cellulose bead that has been surface-modified with covalently attached polylysine chains that have a high affinity for LPS but lacks the toxicity of other endotoxin-binding molecules. With a 2,000,000 EU/ml binding capacity, endotoxin levels can be reduced by 99% in samples containing initial endotoxin levels of 10,000 EU/ml. Typical protein samples processed with the High Capacity Endotoxin Removal Resin have a final endotoxin concentration below 5 EU/ml. This resin is offered in a slurry format for custom packing of endotoxin removal columns for gravity flow or flow rates ranging from 10 to 15 ml/hour.
Delivery information: Supplied as 50% slurry in 20% ethanol
Caution: For Research Use Only. Not for use in diagnostic procedures.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...