Pierce™ HisPur™ Ni-NTA Spin Columns, Thermo Scientific
Supplier: Thermo Scientific
88224 88225 88226
CAPI88224PK 572.13 CAD
CAPI88224 CAPI88225 CAPI88226
Pierce™ HisPur™ Ni-NTA Spin Columns, Thermo Scientific
Filters Centrifugal Filters
Pierce™ HisPur™ Ni-NTA Spin Columns contain a high-capacity, high-performance nickel-IMAC resin for routine affinity purification of His-tagged fusion proteins.
  • High capacity binding up to 60 mg of 6×His-tagged protein per milliliter of resin
  • Purify proteins using native or denaturing conditions
  • Resin can be reused at least five times without reduction in performance
  • Purifies high levels of overexpressed His-tagged fusion proteins from bacterial lysates
The specially prepared support consists of beaded agarose derivatized with the nitrilotriacetic acid (NTA) chelation moiety and loaded with divalent nickel ions (Ni²⁺). The immobilized metal affinity chromatography (IMAC) resin provides exceptional binding capacity and performance for recombinant His-tagged protein purification.
Caution: For Research Use Only. Not for use in diagnostic procedures and it should be store at 4 °C.