Pierce™ HisPur™ Ni-NTA Resin, Thermo Scientific
Supplier: Thermo Scientific
88221 88222 88223
CAPI88221EA 308.28 CAD
CAPI88221 CAPI88222 CAPI88223
Pierce™ HisPur™ Ni-NTA Resin, Thermo Scientific
Protein Purification Protein Purification Resins
Pierce™ HisPur™ Ni-NTA Resin is a high-capacity, high-performance nickel-IMAC resin for routine affinity purification of His-tagged fusion proteins.
  • Binding up to 60 mg of 6xHis-tagged protein per milliliter of resin
  • Purify proteins using native or denaturing conditions
  • Reuse the same batch of resin at least five times
  • Pre-formulated buffers available for kit formats
The specially prepared support consists of beaded agarose derivatized with the nitrilotriacetic acid (NTA) chelation moiety and loaded with divalent nickel ions (Ni²⁺). The immobilized metal affinity chromatography (IMAC) resin provides exceptional binding capacity and performance for recombinant His-tagged protein purification.
Caution: For Research Use Only. Not for use in diagnostic procedures.