Pierce™ Subcellular Protein Fractionation Kit for Tissues, Thermo Scientific
Supplier: Thermo Scientific
87790 87791
CAPI87790EA 1482.12 CAD
CAPI87790 CAPI87791
Pierce™ Subcellular Protein Fractionation Kit for Tissues, Thermo Scientific
Buffers Extraction Buffers
The Pierce™ kit is ideal for cell fractionation and enrichment of proteins from five different cellular compartments within two hours.
  • Optimized - formulations and protocols specific for fractionation of tissue
  • Complete - obtain cytoplasmic, membrane, soluble nuclear, chromatin-bound and cytoskeletal protein fractions from a single kit
  • Complete - obtain protein fractions from a single kit
  • Convenient - unique tissue strainer allows rapid removal of tissue debris
  • Versatile - extract proteins from a variety of tissue types, including liver, heart, brain, kidney, lung and spleen
  • Compatible - use extracts for downstream applications such as protein assays, Western blotting, protein assays, electrophoretic mobility shift assays, reporter gene and enzyme activity assays
Delivery information: Kit includes four extraction buffers, tissue strainers, a stabilized nuclease and halt protease inhibitor cocktail.
Caution: For Research Use Only. Not for use in diagnostic procedures.