Pierce™ Slide-A-Lyzer™ Dialysis Flasks, Thermo Scientific
Supplier: Thermo Scientific
87760 87761 87762 87763
CAPI87760PK 769.07 CAD
CAPI87760 CAPI87761 CAPI87762 CAPI87763
Pierce™ Slide-A-Lyzer™ Dialysis Flasks, Thermo Scientific
Tubing Dialysis Tubing
Pierce™ Slide-A-Lyzer™ Dialysis Flasks facilitate simple and effective removal of buffer salts and small contaminants from proteins and other macromolecules in sample volumes up to 250 ml.
  • Easy to use
  • Color-coded for easy identification
  • No leakage, sample remains intact with no contamination
  • High surface area to volume ratio maximizes diffusion rate
Slide-A-Lyzer™ Dialysis Flasks are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of an inert plastic body.
With Slide-A-Lyzer Dialysis Flasks, typical low-molecular weight contaminant removal, buffer exchange and desalting can be accomplished in 8 hours to overnight.
The membrane and flask materials are compatible with most laboratory buffers.
Caution: For Research Use Only. Not for use in diagnostic procedures.