Pierce™ High Capacity Streptavidin Chromatography Cartridges, Thermo Scientific
Supplier: Thermo Scientific
87739 87740
CAPI87739EA 645.05 CAD
CAPI87739 CAPI87740
Pierce™ High Capacity Streptavidin Chromatography Cartridges, Thermo Scientific
Chromatography Columns
This high capacity Streptavidin chromatography cartridges are convenient, ready-to-use devices for separating biotinylated molecules from non-biotinylated molecules and for purifying antigens using biotinylated antibodies.
  • Enable fast, easy and reproducible chromatographic separations
  • Can be regenerated multiple times for affinity purification
  • Capacity greater than 10 mg biotinylated BSA/mL of resin
  • Contains high-quality resin slurry
  • Product shipped at ambient temperature
  • Upon receipt do not freeze the resin and store at 4 ˚C
These cartridges are compatible with the major automated liquid-chromatography systems or manual syringe processing
Available in several bottle sizes of resin slurry and as packed 1 and 5 ml chromatography cartridges.
The cartridges attach directly to ÄKTA™ or FPLC systems without additional connectors. The included accessory pack readily adapts cartridges for use with Luer-Lok™ syringe fittings or ¹/₁₆" tubing.
Caution: For research use only, not for use in diagnostic procedures.