Halt™ Phosphatase Inhibitor Cocktail, Thermo Scientific
Supplier: Thermo Scientific
78420 78426 78427 78428
CAPI78420EA 569.13 CAD
CAPI78420 CAPI78426 CAPI78427 CAPI78428
Halt™ Phosphatase Inhibitor Cocktail, Thermo Scientific
Cell Culture Supplements and Additives
Thermo Scientific Halt Phosphatase Inhibitor Cocktail preserves the phosphorylation state of proteins during and after cell lysis or tissue protein extraction.
  • All in one format - Halt Phosphatase Inhibitor Cocktail inhibits both serine/threonine as well as protein tyrosine phosphatases
  • Compatible with standard protein assays
  • User friendly storage at 2 to 8 °C - Halt Phosphatase Inhibitor Cocktail does not require frozen storage
  • Convenient size - 1 mL of cocktail protects up to 100 mL of sample
The cocktail contains a mixture of four phosphatase inhibitors of broad specificity, including sodium fluoride, sodium orthovanadate, sodium pyrophosphate and -glycerophosphate. Unlike other commercially available phosphatase inhibitor cocktails, which protect against either serine/threonine phosphatases or protein tyrosine phosphatases, the Halt Phosphatase Inhibitor Cocktail protects phosphoproteins from both families of phosphatases. Conveniently packaged in a 100X format, the cocktail is stable at 4 °C and is compatible with common protein assays, including Bradford-based assays.
Caution: For Research Use Only. Not for use in diagnostic procedures.