Pierce™ DyLight™ Antibody Labeling Kits, Thermo Scientific
Supplier: Thermo Scientific
The DyLight™ Antibody Labeling Kits and were specifically developed for fast, efficient labeling of antibodies
- Fast – fluorescently label and purify protein in approximately one hour
- Amine-reactive dyes – label virtually any protein
- Pre-measured fluorescent dye – eliminate the time, waste and hassle associated with weighing dye
- Efficient non-reacted dye removal
- Minimal sample dilution
- Spin column format eliminates the need for column preparation, fraction screening and waiting for protein to emerge from column
- Easy protocol
Two convenient kit formats are available to accommodate varied labeling requirements. The DyLight™ Antibody Labeling Kits contain all necessary components to perform three separate labeling reactions using 1mg of IgG or similar quantities of other proteins. The DyLight™ Microscale Antibody Labeling Kits contain all the necessary components to perform five separate labeling reactions using 100 μg of IgG. The labeling kits use high-performance spin desalting columns to provide exceptional dye removal and antibody recovery.
Ordering information: Standard kit includes Reaction Buffer (1 mL), spin columns (6 each), microcentrifuge collection tubes (12 each) and purification resin (5 mL). Each Microscale kit includes Reaction Buffer (1 mL), spin columns (5 each), microcentrifuge collection tubes (10 each) and purification resin (5 mL).
Caution: For Research Use Only. Not for use in diagnostic procedures.