Pierce™ Protein A/G and A/G Plus UltraLink™ Resin, Thermo Scientific
Supplier: Thermo Scientific
53132 53133 53135
CAPI53132EA 987.49 CAD
CAPI53132 CAPI53133 CAPI53135
Pierce™ Protein A/G and A/G Plus UltraLink™ Resin, Thermo Scientific
Protein Purification Protein Purification Resins
Pierce™ Protein A/G UltraLink™ and Protein A/G Plus UltraLink™ resins are extremely durable beaded polyacrylamide supports to which recombinant Protein A/G has been immobilized for use in IgG purification methods
  • Specifically binds IgG from several mammalian species, especially most subtypes of human and rabbit IgG
  • Acrylamide-based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications
  • Used to bind and purify IgG in gravity-flow, spin-column or medium-pressure chromatography methods
Immobilized proteins are covalently coupled to an agarose support to provide an efficient method of purifying antibodies.
Caution: For Research Use Only. Not for use in diagnostic procedures.