Pierce™ Streptavidin Plus UltraLink™ Resin, Thermo Scientific
Supplier: Thermo Scientific
Pierce™ Streptavidin Plus UltraLink™ resin consists of purified recombinant streptavidin that has been covalently immobilized onto a beaded acrylamide support
- Acrylamide based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications
- Streptavidin Plus UltraLink™ resin binding capacity: >4 mg of biotinylated BSA/mL of resin
UltraLink™ beads are hydrophilic, charge free, highly crosslinked, rigid and durable which makes the affinity resin suitable for a variety of batch or column type affinity procedures that require medium pressure chromatography.
Caution: For Research Use Only. Not for use in diagnostic procedures.